Learn More
Perkin Elmer US LLC Silver Gate Flat Crystal Top Plate - Ge, Single Reflection
Supplier:  Perkin Elmer US LLC L1272275
Encompass_Preferred
This top plate is used with Silver Gate ATR accessories which are compatible with the Spectrum One, 100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3520