Learn More
Strem, An Ascensus Company CAS# 26042-64-8. 5g. Silver hexafluoroantimonate(V), 98%. MFCD00003401
SDP
Supplier:  Strem, An Ascensus Company 9347485G
Encompass
CAS# 26042-64-8. 5g. Silver hexafluoroantimonate(V), 98%. MFCD00003401. Molecular Weight: 343.61. Molecular Formula: AgSbF6. Color/form: white to off-white pwdr. Strem# 93-4748. http://www.strem.com/catalog/v/93-4748/
Catalog No. 50-901-15434