Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver hydrogen fluoride, 99% min, Thermo Scientific Chemicals

Catalog No. p-7050969
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4012909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4012909 Supplier Thermo Scientific Chemicals Supplier No. 04012909
Only null left
Add to Cart

Silver hydrogen fluoride facilitates the photochemical conversion of carboxylic acids to alkyl fluorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12249-52-4
MDL Number MFCD00042145
Synonym silver hydron fluoride, silver salt, h, fluoride, silver 1+ ion hydrogen fluoride, hydrogen . silver 1+ fluoride

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Linear Formula AgHF2
UN Number UN1740
Solubility Information Soluble in water.
Formula Weight 146.87
Percent Purity ≤99%
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Silver hydrogen fluoride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROGENDIFLUORIDE, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.