Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) Oxide, Solstice

Catalog No. 6001359 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-59 10 g
60-013-61 50 g
60-013-60 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-013-59 Supplier Solstice Supplier No. 22116310G

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Quantity 10 g
UN Number 1479
Formula Weight 231.74g/mol
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material Silver(I) Oxide
Hazard Statement P220-P273-P280-P305 + P351 + P338-P501
Precautionary Statement H271-H318-H410
EINECSNumber 243-957-1
RTECSNumber VW4900000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.