Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) Oxide, purum p.a, ≥99.0% (AT), Solstice

Catalog No. 60047695 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-695 10 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-695 Supplier Solstice Supplier No. 8526010G

missing translation for 'validMainframeMsg'

Purum p.a.,≥99.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Color Gray
Physical Form Powder
Quantity 10 g
UN Number UN1479
Solubility Information Slightly soluble
Formula Weight 231.74g/mol
Percent Purity ≥99.0% (AT)
Odor Odorless
Packaging Poly Bottle
Chemical Name or Material Silver(I) oxide
Hazard Statement P220-P273-P280-P305 + P351 + P338-P501
DOTInformation class 5.1 - PG 1 - EHS - Oxidizing solid, n.o.s., HI: all
RTECSNumber 243-957-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.