Learn More
Strem, An Ascensus Company CAS# 7783-95-1 Silver(II) fluoride, 98% MFCD00003411. 25G
SDP
Catalog No. 5090115432
Encompass
CAS# 7783-95-1. 25G. Silver(II) fluoride, 98%. MFCD00003411. Molecular Formula: AgF2 Molecular Weight: 145.87. Color/Form: brown pwdr. Stability: light sensitive, hygroscopic Strem#: 93-4747 www.strem.com/catalog/v3/93-4747/
Catalog No. 50-901-15432 Supplier Strem, An Ascensus Company Supplier No. 93474725G
CAS# 7783-95-1. 25G. Silver(II) fluoride, 98%. MFCD00003411. Molecular Formula: AgF2 Molecular Weight: 145.87. Color/Form: brown pwdr. Stability: light sensitive, hygroscopic Strem#: 93-4747 www.strem.com/catalog/v3/93-4747/