Learn More
Strem, An Ascensus Company CAS# 1301-96-8. 10g. Silver(II) oxide (99.9%-Ag). MFCD00044285
SDP
Catalog No. 5090115423
Encompass
CAS# 1301-96-8. 10g. Silver(II) oxide (99.9%-Ag). MFCD00044285. Molecular Weight: 123.87. Molecular Formula: AgO. Color/form: black pwdr. Strem# 93-4742. http://www.strem.com/catalog/v/93-4742/
Catalog No. 50-901-15423 Supplier Strem, An Ascensus Company Supplier No. 93474210G
CAS# 1301-96-8. 10g. Silver(II) oxide (99.9%-Ag). MFCD00044285. Molecular Weight: 123.87. Molecular Formula: AgO. Color/form: black pwdr. Strem# 93-4742. http://www.strem.com/catalog/v/93-4742/