Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99% | 7440-22-4 | MFCD00003397 | 1EA
Catalog No. 501853141
Encompass
Silver insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99% | Purity: 99.99% | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 1EA
Catalog No. 50-185-3141 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF793580021EA
Silver insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99% | Purity: 99.99% | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 1EA