Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silver iodide | 7783-96-2 | MFCD00003412 | 99.0% | 234.77 g/mol | AgI | 25 G
SDP

Catalog No. 5000303036
Encompass_Preferred

Silver iodide (AgI) is an inorganic photosensitive compound used as a laboratory reagent in life-science research and materials applications. It is supplied as a high-purity solid with supporting documentation for safe handling and quality control.

  • High purity: 99.0%.
  • Compact pack size: 25 g.
  • CAS number 7783-96-2 for clear identification.
  • Chemical formula AgI; molecular weight 234.77 g/mol.
  • Photosensitive properties useful in photographic and materials research.
  • SDS and COA available to support safety and quality requirements.

Catalog No. 50-003-03036 Supplier Medchemexpress LLC Supplier No. 25GHYW019930
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver iodide (AgI) is an inorganic photosensitive compound used as a laboratory reagent in life-science research and materials applications. It is supplied as a high-purity solid with supporting documentation for safe handling and quality control.

  • High purity: 99.0%.
  • Compact pack size: 25 g.
  • CAS number 7783-96-2 for clear identification.
  • Chemical formula AgI; molecular weight 234.77 g/mol.
  • Photosensitive properties useful in photographic and materials research.
  • SDS and COA available to support safety and quality requirements.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.