Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver iodide, 99.999%, (trace metal basis), extra pure, Thermo Scientific™

Catalog No. AC194060500
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC194060500 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC194060500 Supplier Thermo Scientific Chemicals Supplier No. 194060500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-96-2
Quantity 50g
Assay Percent Range 100.00%
Molecular Formula AgI
Linear Formula AgI
MDL Number MFCD00003412
Merck Index 15, 8655
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES [Ag]I
IUPAC Name iodosilver
Molecular Weight (g/mol) 234.773
PubChem CID 24563
Formula Weight 234.77
Grade Trace Metal
Density 5.68g/mL
Chemical Name or Material Silver iodide, 99.999%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Avoid release to the environment.
EINECSNumber 232-038-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.