Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver lactate, 96%

Catalog No. AC233330050
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233330050 5 g Glass bottle
AC233330100 10 g Glass bottle
AC233330500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC233330050 Supplier Thermo Scientific Chemicals Supplier No. 233330050

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

missing translation for 'validMainframeMsg'

CAS: 15768-18- | C3H5O3Ag | 196.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15768-18-
Molecular Formula C3H5O3Ag
Molecular Weight (g/mol) 196.94
InChI Key LMEWRZSPCQHBOB-UHFFFAOYSA-M
Synonym silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
PubChem CID 91824762
IUPAC Name silver;2-hydroxypropanoate
SMILES CC(C(=O)[O-])O.[Ag+]

Specifications

Melting Point 120.0°C to 122.0°C
Color Brown-Gray to Gray
Physical Form Powder
Quantity 5 g
Assay Percent Range 95% min. (Argentometry)
Linear Formula CH3CH(OH)CO2Ag
Merck Index 14,8517
Solubility Information Solubility in water: 77g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 196.94
Percent Purity 96%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Silver lactate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protectiv
EINECSNumber 239-859-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.