Learn More
Electron Microscopy Sciences Silver Lactate, Powder 5 GR
SDP
Catalog No. 5028618
Encompass_Preferred
CAS #128-00-7 CH₃CH(OH)CO₂Ag F.W. 196.94
Silver content is approximately 90% min. Used to localize intracellular anions (Cl-).
Catalog No. 50-286-18 Supplier Electron Microscopy Sciences Supplier No. 21000
CAS #128-00-7 CH₃CH(OH)CO₂Ag F.W. 196.94
Silver content is approximately 90% min. Used to localize intracellular anions (Cl-).