Learn More
Sterlitech Corporation Sterlitech Silver Membrane, 0.2 Micron, 47mm, 25/Pk
SDP
Supplier:  Sterlitech Corporation 45348
Encompass_Preferred
Pure metallic silver (99.97% pure silver); inorganic, hydrophilic membrane filter; pore rating of 0.2 microns and a diameter of 47 mm.
Catalog No. NC1135913