Learn More
Sterlitech Corporation Sterlitech Silver Membrane, 0.45 Micron, 47mm, 25/Pk
SDP
Supplier:  Sterlitech Corporation 45347
Encompass_Preferred
Pure metallic silver (99.97% pure silver), hydrophilic membrane filter, pore rating of 0.45 microns and a diameter of 47 mm
Catalog No. NC1360772