Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver nanoparticles ink for inkjet printing

Catalog No. 90108325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-108-325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-108-325ML Supplier Sigma Aldrich Supplier No. 90108325ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Smart'Ink S-CS01130 conductive ink was developed for the printed electronics, is particularly well suited for applications requiring high conductivity and high resolution. This ink, based on silver, is perfectly adapted to design conductive traces on flexible substrates. Also, it is shaped for IoT application which required antennas (HF, UHF).Smart'Ink S-CS01130 conductive ink shows improved jetting performance and is successfully used on different tools such as Drop on Demand (DOD) inkjet printer FUJIFILM Dimatix, or such as digital micro-dispensing inkjet printer Microdrop and Microfab.Product of Genes'Ink.

Specifications

Linear Formula Ag
Quantity 25 mL
Synonym Ag ink; Conductive ink; SmartInk S-CS01130 conductive ink
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.