Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.0282 N, 1 mL = 1 mg Cl-, Ricca Chemical
SDP

Catalog No. 69501 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6950-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6950-1 Supplier Ricca Chemical Company Supplier No. 69501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7761-88-8
Molecular Formula AgNO3
MDL Number MFCD00003414
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
Equivalence 1 mL = 1 mg Cl
PubChem CID 24470
ChEBI CHEBI:32130
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.282N
Packaging Black Poly Bottle
pH ∼6
Quantity 4 L
Chemical Name or Material Silver Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.