Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.1 M Solution, Innovating Science™
SCI_ED

Catalog No. S000227 Shop All Aldon Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000227 100 mL
S000228 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000227 Supplier Aldon Corporation Supplier No. IS28038
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 100°C
Chemical Name or Material Silver Nitrate
Color Colorless
CAS 7732-18-5, 7761-88-8
Physical Form Liquid
Quantity 100 mL
Synonym Silver(I) nitrate, Water solution
Solubility Information Soluble in Water
PubChem CID 24470
Grade Reagent
Concentration or Composition (by Analyte or Components) 0.1 M
Odor Odorless
Packaging Amber Glass
Appearance Colorless Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.