Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.171 N Standardized Solution, Spectrum™ Chemical
SDP

Catalog No. 18613179 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-179 4 L Glass Bottle
18-613-177 500 mL Amber Glass Bottle
18-613-180 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-613-179 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1664LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Quantity 4 L
Linear Formula AgNO3
Synonym Lunar caustic, Nitric acid silver salt
Solubility Information Miscible in Water
PubChem CID 24470
Formula Weight 169.87 g/mol
Concentration or Composition (by Analyte or Components) 0.171 N ± 0.005 N
Packaging Glass Bottle
Appearance Colorless Liquid
Recommended Storage Light Sensitive, Keep tightly closed in light-resistant containers

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.