Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrate, 0.1N Standardized Solution

Catalog No. p-4407815
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35601AP 500 mL
AA35601K2 1 L
AA35601K7 4 L
AA35601M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA35601AP Supplier Thermo Scientific Chemicals Supplier No. 035601AP
Only null left
Add to Cart

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
Solubility Information Miscible with water,alcohol and aqueous ammonia.
Formula Weight 169.87
Concentration or Composition (by Analyte or Components) Silver nitrate: 1.7%; Water: 98.3%
Odor Odorless
Sensitivity Light sensitive
Appearance Colourless liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Silver nitrate
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 231-853-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.