Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.282 Normal, Reagents

Catalog No. 01333590
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-333-590 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-590 Supplier Reagents Holdings Llc Supplier No. CS1508004C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver Nitrate, 0.282 Normal 1 mL = 10 mg Cl, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 6
Physical Form Liquid
Quantity 4 L
Linear Formula AgNO3
Solubility Information Soluble in Water
PubChem CID 24470
Formula Weight 169.87 g/mol
Grade Reagent
Odor Odorless
Packaging Glass Bottle
Appearance Colorless Liquid
Specific Gravity 1.0401
Chemical Name or Material Silver Nitrate
Show More Show Less
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.