Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 1.00 Normal, Reagents

Catalog No. 01333668
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-668 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-668 Supplier Reagents Holdings Llc Supplier No. CS1400001B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver Nitrate, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless
pH 6
Physical Form Liquid
Quantity 1 L
Linear Formula AgNO3
Solubility Information Miscible in Water
PubChem CID 24470
Formula Weight 169.87 g/mol
Grade Reagent
Odor Odorless
Packaging Opaque HDPE
Appearance Colorless Liquid
Specific Gravity 1.1329
Chemical Name or Material Silver Nitrate, 1.00 Normal
Show More Show Less
ShelfLife 2 Years
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.