Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 1.7% (w/v) (17 g/L), Ricca Chemical
SDP

Catalog No. 6825716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
68257-16 500 mL Amber Poly Bottle
68257-32 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68257-16 Supplier Ricca Chemical Company Supplier No. 6825.716
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Min % 96.0, 1.64
CAS Max % 100.0, 1.71
Physical Form Liquid
Quantity 500 mL
Percent Purity 1.7% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1.7% (w/v) Aqueous Solution (17g/L)
Packaging Amber Poly Bottle
Chemical Name or Material Silver Nitrate
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.