Learn More
Electron Microscopy Sciences Silver Nitrate 1% Aqueous Solution 250 ML
SDP
Catalog No. 5030039
Encompass_Preferred
Used in Modified Steiner – Using Chapman's Modification for bacterial stain. Spirochetes, Donovan Bodies, General. See references: Margeson L.S. Chapman CM: Use of Zinc Formalin as a sensitizer in Silver Stain for pirochetes. Journal of Histology 19:2, p 135- 138, 6 –96 (1996)
Catalog No. 50-300-39 Supplier Electron Microscopy Sciences Supplier No. 2611902
Used in Modified Steiner – Using Chapman's Modification for bacterial stain. Spirochetes, Donovan Bodies, General. See references: Margeson L.S. Chapman CM: Use of Zinc Formalin as a sensitizer in Silver Stain for pirochetes. Journal of Histology 19:2, p 135- 138, 6 –96 (1996)