Learn More
Electron Microscopy Sciences Silver Nitrate 5% 250 ML
SDP
Catalog No. 5031872
Encompass_Preferred
Used in Gomori's Methenamine Silver for Urate Crystals Urate crystals. Sheehan, Dezna C. and Hrapchak, Barbara B.: Theory and Practice of Histotechnology, 2nd Ed. The C.V. Mosby Company, St. Louis, p.225-226, c. 1980.
Catalog No. 50-318-72 Supplier Electron Microscopy Sciences Supplier No. 2661401
Used in Gomori's Methenamine Silver for Urate Crystals Urate crystals. Sheehan, Dezna C. and Hrapchak, Barbara B.: Theory and Practice of Histotechnology, 2nd Ed. The C.V. Mosby Company, St. Louis, p.225-226, c. 1980.