Learn More
Electron Microscopy Sciences Silver Nitrate 5% Solution 250 ML
SDP
Catalog No. 5030064
Encompass_Preferred
(Kernechtrot)
For bone and its related stain. See references Pizzolato, P., J. Histochem. Cytochem., 12:333-136, 1964. and Luna, L.G., (ed), Manual of Staining Methods of the AFIP, 3rd Ed, McGraw-Hill, NY, c 1968, p 77.
Catalog No. 50-300-64 Supplier Electron Microscopy Sciences Supplier No. 2621201
(Kernechtrot)
For bone and its related stain. See references Pizzolato, P., J. Histochem. Cytochem., 12:333-136, 1964. and Luna, L.G., (ed), Manual of Staining Methods of the AFIP, 3rd Ed, McGraw-Hill, NY, c 1968, p 77.