Learn More
Strem, An Ascensus Company Silver nitrate 3N (ACS) 500g
SDP
Catalog No. 5090115402
Encompass
CAS# 7761-88-8. 500g. Silver nitrate (99.9%-Ag) (ACS). MFCD00003414. Molecular Weight: 169.87. Molecular Formula: AgNO3. Color/form: white xtl. Strem# 93-4718. http://www.strem.com/catalog/v/93-4718/
Catalog No. 50-901-15402 Supplier Strem, An Ascensus Company Supplier No. 934718500G
CAS# 7761-88-8. 500g. Silver nitrate (99.9%-Ag) (ACS). MFCD00003414. Molecular Weight: 169.87. Molecular Formula: AgNO3. Color/form: white xtl. Strem# 93-4718. http://www.strem.com/catalog/v/93-4718/