Learn More
Newcomer Supply Silver Nitrate, ACS Powder, 25 g
SDP
Catalog No. NC2101928
Encompass
Shelf Life is 4 years from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101928 Supplier Newcomer Supply Supplier No. 13803A
Shelf Life is 4 years from date of manufacture. Store at 15-30°C.