Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrate, ACS Reagent, ≥99.0%, Honeywell Fluka™

Catalog No. p-7121628 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-182 25 g Glass Bottle
60-045-181 100 g Glass Bottle
60-013-18 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-182 Supplier Honeywell Chemicals Supplier No. 20913925G

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C (decomposition)(literature)
CAS Max % ≤100.0000%
Quantity 25 g
Linear Formula AgNO3
UN Number UN1493
IUPAC Name silver;nitrate
Formula Weight 169.87
Percent Purity >99%
Grade ACS Reagent
Packaging Glass Bottle
Density 4.350 g/cm3; Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate, ACS Reagent, ≥99.0%
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314-H410
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 5.1; Packing Group:3: II; Proper Shipping Name: Silver nitrate
EINECSNumber 231-853-9
RTECSNumber VW4725000
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.