Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
30g
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7761-88-8
AgNO3
169.872
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
CHEBI:32130

High quality chemicals for laboratory and specialized industrial use

SMILES [N+](=O)([O-])[O-].[Ag+]
IUPAC Name silver;nitrate
Formula Weight 169.87
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.35kg
Quantity 30g
Chemical Name or Material Silver Nitrate
Grade Baker Analyzed™ ACS Reagent