Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. p-7158165 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-231 25 g Amber Glass Bottle
18-606-232 100 g Amber Glass Bottle
18-606-233 125 g Amber Glass Bottle
18-606-234 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-231 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S108525GM
Only null left
Add to Cart
Add to Cart

S1085, 7761-88-8, AgNO3

Spectrum™ Chemical Silver Nitrate, Crystal, Reagent, ACS is an inorganic compound with the formula AgNO3. Silver nitrate is an extremely versatile analytical laboratory reagent for both the qualitative and quantitative determination of halides, carbonates, hydroxides, sulfides and phosphates. Spectrum provides solid Reagent ACS grade silver nitrate for preparation of analytical laboratory reagent solutions. As an ACS grade Reagent, Spectrum Chemical manufactured Silver Nitrate is used as the quality reagent in standard testing to which other substances are graded. It has met the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.