Learn More
Silver Nitrate, Crystal, USP, 99.8-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| Boiling Point | 440°C |
| Melting Point | 212°C |
| Color | White |
| CAS Max % | 1 |
| Physical Form | Fine Crystals or Crystalline Powder |
| Quantity | 100 g |
| Linear Formula | AgNO3 |
| Synonym | Lunar caustic, Caustic silver, Silver(1+) nitrate |
| Solubility Information | Soluble in Water |
| PubChem CID | 24470 |
| Show More |
Safety and Handling
| Recommended Storage | Light Sensitive, Keep tightly closed in light-resistant containers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.