Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, Crystals, GR, ACS, MilliporeSigma™

Catalog No. MSX02055 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
125 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX02055 125 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX02055 Supplier MilliporeSigma Supplier No. SX02055

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444°C (1013hPa)(decomposition)
Melting Point 212°C
Color Colorless
pH 5.4 to 6.4 (100g/L, H2O, 20°C)
Physical Form Crystals
Quantity 125 g
UN Number UN 1493
Solubility Information 2160 g/L
Formula Weight 169.87
Percent Purity 99.0%
Odor Odorless
Packaging Glass Bottle
Density 4.35 g/cc at 20°C; Bulk Density: ∼2350 kg/m3
Chemical Name or Material Silver Nitrate
Show More Show Less
Hazard Category Danger
Hazard Statement Hazard StatementsH272 May intensify fire; oxidizer. H314 Causes severe skin burns and eye damage.
Precautionary Statement Precautionary StatementsP210 Keep away from heat. P220 Keep/Store away from clothing/ combustible materials. P221 Take any precaution to avoid mixing with combustibles. P260 Do not breathe dusts or mists. P264 Wash skin thoroughly after handling. P280 Wear protective gloves/ protective clothing/ eye protection/ face protection. P301 + P330 + P331 IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. P303 + P361 + P353 IF ON SKIN (or hair): Remove/ Take off immediately all contaminated clothing. Rinse skin with water/ shower. P304 + P340 IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P310 Immediately call a POISON CENTER or doctor/ physician. P321 Specific treatment (see supplemental first aid instructions on this label). P363 Wash contaminated clothing before reuse. P370 + P378 In case of fire: Use dry sand, dry chemical or alcohol-resistant foam for extinction. P405 Store locked up. P501 Dispose of contents/ container to an approved waste disposal plant
EINECSNumber 231-853-9
RTECSNumber VW4725000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.