Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.00500 N (N/200) in Isopropyl Alcohol, Ricca Chemical
SDP

Catalog No. R70258001B Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R70258001B 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R70258001B Supplier Ricca Chemical Company Supplier No. R70258001B
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0 , 7732-18-5 , 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Physical Form Solution
Quantity 1 L
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Characteristic alcohol
Packaging Amber Poly Bottle
Appearance Clear colorless to slight tan liquid
Chemical Name or Material Silver Nitrate
Recommended Storage Flammable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.