Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001321 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-21 250 g
60-013-20 1 kg
60-013-22 25 g
60-013-19 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-013-21 Supplier Solstice Supplier No. 31630250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212 °C
Color Colorless
Quantity 250 g
Linear Formula AgNO3
UN Number 1493
Solubility Information Soluble in Water
Formula Weight 169.87g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Odor Odorless
Packaging Plastic Bottle
Appearance White Crystals
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314-H410
EINECSNumber 231-853-9
RTECSNumber VW4725000
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.