Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrate solution, Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl), Solstice

Catalog No. 60046580 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-580 6 x 1 L
60-013-29 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-580 Supplier Solstice Supplier No. 353776X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 100°C
CAS Min % 99.0000%
CAS Max % 100.0000%, <1.0000%
Quantity 6 x 1 L
Linear Formula AgNO3
Solubility Information Soluble in Water
IUPAC Name silver;nitrate
Formula Weight 169.87
Grade Volumetric
Concentration or Composition (by Analyte or Components) 0.0282 M AgNO3 (1 ml = 1 mg Cl)
Packaging Glass Bottle
Appearance Colorless Liquid
Density 1.000 g/cm3
Chemical Name or Material Silver nitrate solution, Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl)
Show More Show Less
ShelfLife 1080 days from date of manufacture
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.