Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate Solution (0.1 N (N/10)/Certified), Fisher Chemical PROMO

Catalog No. SS724FB25 Shop All Fisherbrand Products
Change view
Click to view available options
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SS724FB25 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SS724FB25

missing translation for 'validMainframeMsg'

Standardized at 25°C against NIST™ potassium chloride.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87 g/mol
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
UN Number 3082
Concentration 0.0995 to 0.1005 N
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Traceability to NIST 999b POTASSIUM CHLORIDE
Vapor Pressure 14 mmHg at 20°C
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Silver Nitrate Solution, 0.1 N (N/10)
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement

Emergency Overview: May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

NFPA

Health:1

Flammability:0

Instability:0

DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.