Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Silver Nitrate Solution, 0.5M
SCI_ED

Catalog No. S25874 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25874 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25874
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepare for qualitative analysis and inorganic reactions with silver nitrate solution. Ideal for instructional chemistry labs.

  • Chemical Name or Material: Silver Nitrate Solution
  • Concentration: 0.5 Molar (0.5M)
  • Quantity: 500 mL
  • CAS Numbers: 7761-88-8 (Silver Nitrate), 7732-18-5 (Water)
  • CAS Max Percent: Silver Nitrate: 10.0% Water: 90.0%
  • Molecular Weight: 169.87 g/mol
  • Synonyms: Silver nitrate, Lunar caustic, Silbernitrat, Argenti nitras, Silver mononitrate, Nitric acid silver(I) salt
  • For Use In: Educational settings including high school and college chemistry labs
  • Product Use: Ideal for studying precipitation reactions, qualitative analysis, and ionic compound interactions

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5 , 7761-88-8 , 7732-18-5 , 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

CAS Min % 0.0
CAS Max % 10.0, 90.0
Density 1.000
Grade Reagent
Packaging 0.5M
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Silver Nitrate Solution, 0.5M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.