Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate Solution, 1.0M, Solstice

Catalog No. 6001337 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-37 500 mL
60-013-36 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-013-37 Supplier Solstice Supplier No. 319449500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 1.0 M AgNO3 (1.0N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Quantity 500 mL
Linear Formula AgNO3
UN Number 3264
Solubility Information Miscible in Water
Formula Weight 169.87g/mol
Grade Volumetric
Concentration or Composition (by Analyte or Components) 1.0M
Odor Odorless
Packaging Glass Bottle
Appearance Colorless Liquid
Density 1.135g/mL at 25°C; Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate Solution
Show More Show Less
Hazard Statement P220-P273-P280-P305 + P351 + P338-P310-P501
Precautionary Statement H314-H410
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.