Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Silver Nitrate Solution (Fontana) - a - Poly Scientific Special Stain Kit Component
SDP

Catalog No. 23640212
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-212 8 oz.
23-640-211 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-212 Supplier StatLab Supplier No. S181S8OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Silver Nitrate Solution (Fontana)

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Silver Nitrate solution is used with Fontana Masson Silver Method for Argentaffin Granules
  • Argentaffin substances are able to bind and reduce silver to the visible form

Specifications

Physical Form Aqueous Solution
Color Purple/Violet
Quantity 8 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.