Learn More
Hach Company Silver Nitrate Digital Titrator Cartridge, 0.2256 N
Supplier:  Hach Company 1439601
Encompass
For chloride determination by Digital Titrator argentometric titration. Hach method 8207. Digits x 0.1 = mg/L Cl- in 100 mL sample. Approximately 40-100 tests
Catalog No. 50-199-5018