Learn More
Hach Company 1
Supplier:  Hach Company 31653
Encompass
Silver Nitrate Titrant for chloride determination by buret titration. Hach method 8225. Uses Chloride 2 Indicator Powder Pillows (Cat. No. 105766) to signal the endpoint. 1 Liter Bottle.
Catalog No. 50-199-4246