Learn More
Hach Company Silver Nitrate Digital Titrator Cartridge, 1.128 N
Supplier:  Hach Company 1439701
Encompass
For chloride determination by Digital Titrator argentometric titration. Hach method 8207. Digits = mg/L Cl- in 50 mL sample. Approximately 40-100 tests
Catalog No. 50-199-5019