Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, Technical, Spectrum™ Chemical
SDP

Catalog No. 18613205 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-205 500 g Amber Glass Bottle
18-613-204 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-613-205 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1086500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7761-88-8

Specifications

Boiling Point 440°C
Melting Point 212°C
Color White
CAS Max % 1
Quantity 500 g
Molecular Formula AgNO3
Linear Formula AgNO3
Synonym Argenti nitras, Argerol, Lunar caustic
Solubility Information Solubility in Water, Soluble in Diethyl Ether, Soluble in Alcohol, Soluble in Acetone
InChI Key SDXLMBHZGTSTSA-UHFFFAOYSA-N
IUPAC Name Silver(1+) nitrate
PubChem CID 24470
Formula Weight 169.87 g/mol
Grade Technical
Odor Odorless
Packaging Amber Glass Bottle
Appearance Colorless or White Crystals
Density 4.35 g/cm³
Show More Show Less
Recommended Storage Light Sensitive, Keep tightly closed in light-resistant containers

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.