Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions SILVER NITRATE W-V 10%/100ML
SDP

Catalog No. NC9832632
Encompass_Preferred

Silver Nitrate 10% W/v Aqueous Solution (100ml)

Catalog No. NC9832632 Supplier Aqua Solutions Supplier No. 8442100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver Nitrate 10% W/v Aqueous Solution (100ml)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.