Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silver nitrite | 7783-99-5 | MFCD00003413 | 99.9% | 153.874 g/mol | AgNO2 | 5 G
SDP

Catalog No. 5000299376
Encompass_Preferred

Silver nitrite (AgNO2) is an inorganic reagent used in biochemical assays, antimicrobial research, and synthetic chemistry. It serves as a nitrite source and has been reported to inhibit cell division and bacterial growth by interfering with protein synthesis. Supplied for laboratory research use only, the material is provided in small pack sizes for analytical and experimental applications.

  • Chemical formula: AgNO2.
  • CAS number: 7783-99-5.
  • High purity suitable for analytical and research use.
  • Available in small pack sizes for lab-scale experiments.
  • Intended for research use only, not for clinical or human use.
  • Common applications include biochemical assays, antimicrobial studies, and synthetic chemistry.

Catalog No. 50-002-99376 Supplier Medchemexpress LLC Supplier No. HYW0340675G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver nitrite (AgNO2) is an inorganic reagent used in biochemical assays, antimicrobial research, and synthetic chemistry. It serves as a nitrite source and has been reported to inhibit cell division and bacterial growth by interfering with protein synthesis. Supplied for laboratory research use only, the material is provided in small pack sizes for analytical and experimental applications.

  • Chemical formula: AgNO2.
  • CAS number: 7783-99-5.
  • High purity suitable for analytical and research use.
  • Available in small pack sizes for lab-scale experiments.
  • Intended for research use only, not for clinical or human use.
  • Common applications include biochemical assays, antimicrobial studies, and synthetic chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.