Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrite, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7783-99-5
AgNO2
153.88
MFCD00003413
KKKDGYXNGYJJRX-UHFFFAOYSA-M
silver nitrite,unii-t3mz57ogif,t3mz57ogif,silbernitrit,nitrous acid, silver 1+ salt,silver i nitrite,silvernitrite,acmc-20ajq7,ag.no2,silver nitrite 10g
160904
silver;nitrite
N(=O)[O-].[Ag+]
7783-99-5
AgNO2
153.88
MFCD00003413
KKKDGYXNGYJJRX-UHFFFAOYSA-M
silver nitrite,unii-t3mz57ogif,t3mz57ogif,silbernitrit,nitrous acid, silver 1+ salt,silver i nitrite,silvernitrite,acmc-20ajq7,ag.no2,silver nitrite 10g
160904
silver;nitrite
N(=O)[O-].[Ag+]
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AgNO2
Fieser 01,1011; 06,515; 11,467
Merck Index 15, 8658
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: decomposes in diluted acids,insoluble in alcohol
Formula Weight 153.88
Physical Form Powder and Agglomerates
Percent Purity 99%
Density 4.4500g/mL
Packaging Glass bottle
Solubility Solubility in water: 3.3g/L (20°C). Other solubilities: decomposes in diluted acids, insoluble in alcohol
Specific Gravity 4.45
Color Yellow to Green
Melting Point 140.0°C
Quantity 10g
Chemical Name or Material Silver nitrite
Show More Show Less

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to th

GHS Signal Word: Danger

EINECSNumber : 232-041-7