Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrite, 99%

Catalog No. p-3756701
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210690100 10 g Glass bottle
AC210690500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC210690100 Supplier Thermo Scientific Chemicals Supplier No. 210690100

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 7783-99-5 | AgNO2 | 153.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-99-5
Molecular Formula AgNO2
Molecular Weight (g/mol) 153.88
MDL Number MFCD00003413
InChI Key KKKDGYXNGYJJRX-UHFFFAOYSA-M
Synonym silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
PubChem CID 160904
IUPAC Name silver;nitrite
SMILES N(=O)[O-].[Ag+]

Specifications

Melting Point 140.0°C
Color Yellow to Green
Physical Form Powder and/or Agglomerates
Quantity 10 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula AgNO2
Fieser 01,1011; 06,515; 11,467
Merck Index 15, 8658
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: decomposes in diluted acids, insoluble in alcohol
Formula Weight 153.88
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 4.45
Density 4.4500g/mL
Chemical Name or Material Silver nitrite
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to th
EINECSNumber 232-041-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.