Learn More
Silver Oxide, Powder, 99.9%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Specifications
Specifications
| Color | Black to Brown |
| CAS Max % | 1 |
| Quantity | 25 g |
| Molecular Formula | Ag2O |
| Linear Formula | Ag2O |
| Synonym | Argentous oxide, Divasil, Silver monooxide, Silver monoxide, Silver monoxide (AgO) |
| InChI Key | QWVPASVCHGPNPD-UHFFFAOYSA-N |
| IUPAC Name | Silver(1+) oxide |
| PubChem CID | 11186 |
| Formula Weight | 231.74 g/mol |
| Show More |
Safety and Handling
| Recommended Storage | Light Sensitive, Keep tightly closed in light-resistant containers |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.