Learn More
Structure Probe Inc SILVER PASTE 30G
SDP
Catalog No. NC9245685
Encompass
SPI Conductive Silver Paste Plus, Plus is a high purity, Uniform, Viscous silver particle suspension, Ideal for SEM use, Especially for the mounting of porous samples, 30g
Catalog No. NC9245685 Supplier Structure Probe Inc Supplier No. 05063AB
SPI Conductive Silver Paste Plus, Plus is a high purity, Uniform, Viscous silver particle suspension, Ideal for SEM use, Especially for the mounting of porous samples, 30g