Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver perchlorate hydrate, 99%

Catalog No. p-3777050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210210050 5 g Glass bottle
AC210210250 25 g Glass bottle
AC210211000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC210210050 Supplier Thermo Scientific Chemicals Supplier No. 210210050
Only null left
Add to Cart

CAS: 14242-05-8 | AgClO4 | 207.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14242-05-8
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.31
MDL Number MFCD00149128
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
PubChem CID 15959344
SMILES [Ag+].[O-][Cl](=O)(=O)=O

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Linear Formula AgClO4·xH2O
Merck Index 10, 8358
Solubility Information Solubility in water: 557g/100mL. Other solubilities: soluble in many organic solvents
IUPAC Name silver;perchlorate;hydrate
Formula Weight 207.32
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Silver perchlorate hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.